Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SGTB Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524998

Catalog No. PIPA524998

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with rat based on sequence homology.

SGTB is co-chaperone that binds directly to HSC70 and HSP70 and regulates their ATPase activity.
Specifications

Specifications

SGTB
Polyclonal
PBS with 0.09% sodium azide
Q96EQ0, Q8VD33
SGT2, beta-SGT, small glutamine rich protein with tetratricopeptide repeats 2, small glutamine-rich protein with tetratricopeptide repeats 2, small glutamine-rich tetratricopeptide repeat-containing protein beta
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
218544, 54557
Immunohistochemistry, Western Blot
Unconjugated
SGTB
Liquid
SGTB
KLH conjugated synthetic peptide between 71-100 amino acids from the N-terminal region of human SGTB
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit