Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SH3BGRL3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526310
Description
This antibody is predicted to react with bovine based on sequence homology.
SH3BGRL3 could act as a modulator of glutaredoxin biological activity.Specifications
SH3BGRL3 | |
Polyclonal | |
SH3BGRL3 | |
SH3BP-1, TIP-B1, SH3 domain-binding glutamic acid-rich-like protein 3, SH3 domain-binding protein 1, SH3BGRL3-like protein, TNF inhibitory protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
73723, 83442 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Q9H299, Q91VW3 | |
SH3BGRL3 | |
KLH conjugated synthetic peptide between 24-52 amino acids from the central region of human SH3BGRL3 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction