Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SH3GL1, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA127869

Catalog No. PIPA127869

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA1-27869 detects SH3GL1 from human, mouse, and rat samples.PA1-27869 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The immunogen is a synthetic peptide corresponding to human SH3GL1 protein.
Specifications

Specifications

SH3GL1
Polyclonal
PBS with no preservative
Q99961, Q62419, O35964
CNSA1, EEN, MGC111371, SH3D2B, SH3P8
Chicken
IgY
50μg
Store at -20°C
Primary
6455
Western Blot
Unconjugated
SH3GL1
Purified
SH3GL1
Synthetic peptide corresponding to human SH3GL1 protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit