Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SH3GL1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA522292

Catalog No. PIPA522292


Add to cart

Description

Description

PA5-22292 targets SH3GL1 in ICC, IF, IHC (P), and WB applications and shows reactivity with Human samples. The PA5-22292 immunogen is a recombit fragment corresponding to a region within amino acids 152 and 368 of EEN (Uniprot IDNo.Q99961). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

SH3GL1
Polyclonal
Unconjugated
SH3GL1
Liquid
SH3GL1
Recombit fragment corresponding to a region within amino acids 152 and 368 of EEN (Uniprot IDNo.Q99961)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Q99961
CNSA1, EEN, SH3D2B, SH3P8, EEN fusion partner of MLL, SH3 domain protein 2B, SH3 domain-containing GRB2-like protein 1, SH3-containing Grb-2-like 1 protein, endophilin-2, endophilin-A2, extra 11-19 leukemia fusion, extra eleven-nineteen leukemia fusion gene protein
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
6455
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit