Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHANK1 Rabbit anti-Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14179

Catalog No. PA14179


Add to cart

Description

Description

PA1-4179 detects SHANK1a in rat samples. PA1-4179 has been successfully used in Western blot procedures. By Western blot this antibody detects a ~212 kDa protein representing SHANK1a in rat brain lysate. The PA1-4179 immunogen is a synthetic peptide corresponding to the N-terminal residues E(63) R L L R E Y P Q S F E K G V(77) of rat Shank1a.

Shank is a novel family of postsynaptic density proteins (PSD) that bind to PSD-95-associated protein GKAP. Its putative function is to act as a scaffold protein in the PSD to cross-link NMDA receptors/PSD-95 complexes and couple them to regulators of the actin cytoskeleton. Shank1a is a splice variant of Shank1.
Specifications

Specifications

SHANK1
Polyclonal
Unconjugated
Shank1
Liquid
SHANK1
Synthetic peptide corresponding to the N-terminal residues E(63) R L L R E Y P Q S F E K G V(77) of rat Shank1a.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Rat
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q9WV48
Shank1a, Spank1, Sstrip
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
78957
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit