Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-SHANK3, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232085

Catalog No. NBP232085


Add to cart

Description

Description

SHANK3 Polyclonal antibody specifically detects SHANK3 in Human samples. It is validated for Immunoprecipitation.
Specifications

Specifications

SHANK3
1.0 mg/mL
Immunoprecipitation 2 - 10 ug/mg lysate
Q9BYB0
DEL22q13.3, KIAA1650proline rich synapse associated protein 2, Proline-rich synapse-associated protein 2, ProSAP2, PSAP2SH3 and multiple ankyrin repeat domains protein 3, SH3 and multiple ankyrin repeat domains 3, shank postsynaptic density protein, Shank3, SPANK-2
Rabbit
IgG
100 ul
Store at 4C. Do not freeze.
Polyclonal
Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Burkholderiales, Corynebacteriaceae, White-tufted-ear marmoset and Ralstonia metallidurans (strain CH34 / ATCC 43123 / DSM 2839).
Immunoprecipitation
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
SHANK3
The immunogen this antibody was made to, maps to a region between residue 1691 to 1741 of human SH3 and Multiple Ankyrin Repeat Domains 3 using the numbering given in entry Q9BYB0 (GeneID 85358).
Immunogen affinity purified
RUO
Primary
85358
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only