Learn More
MilliporeSigma™ anti-Shc, Polyclonal
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma™ ST103350UG
Catalog No. ST103350UG
Add to cart
Description
Description
Specifically detects Shc Clone: in Human, Mouse, Rat samples, and it is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation
Shc is an SH2-PTB domain adaptor protein that links various tyrosine kinases to Ras by recruiting the Grb2/SOS complex to the plasma membrane. Note that the~66kDa isoform is not typically expressed in hematapoetic cells.
Specifications
Specifications
Shc
Unconjugated
Rabbit
IgG
RUO
Primary
Recognizes the ∽46, ∽55, and ∽66kDa isoforms of Shc in EGF-treated A431 cells.
ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation
purified
a recombinant protein consisting of the Sh2 domain of human Shc, fused to GST
50μg
−20°C
Polyclonal
Human, Murine, Rat