Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHC Rabbit anti-Human, Mouse, Pig, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 06203
Description
Anti-SHC Antibody is an antibody against SHC for use in immunohistochemistry, immunoprecipitation and western blotting.Specifications
SHC | |
Polyclonal | |
Immunoaffinity Purified immunoglobulin in 0.2 Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30% | |
Rabbit | |
Protein A purfied | |
Primary | |
Human, Mouse, Porcine, Rat | |
Purified |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
SHC1; p66SHC; SHCA; p66; SHC; FLJ26504; p52SHC | |
Recombinant GST fusion protein corresponding to amino acids 366-473 of human SHC-SH2 | |
250 μg | |
NM_003029 | |
−20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction