Learn More
Enzo Life SciencesSupplier Diversity Partner Mouse anti-sheep/goat IgG monoclonal antibody (GT-34) (HRP conjugate) (500 µl)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADISAB400I
Clone: GT-34. Host: Mouse. Isotype: IgG. Immunogen: Purified goat IgG (whole molecule). Species Reactivity: Sheep, Goat. Applications: ELISA, IHC, WB. Recommended dilutions: ELISA (1:25,000). Immunohistochemistry (1:100). Western Blot (1:10,000). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Immunoaffinity purified. Formulation: Liquid. In PBS, pH 7.3, containing 1% BSA and 0.1% 2-chloroacetamide.
Catalog No. 50-667-390