Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-SHEEP IGG (GAMMA) AB AP  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52200318
Affinity purified polyclonal secondary antibody against Sheep IgG, heavy chains (gamma) made in Rabbit and labeled with alkaline phosphatase. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-670-20