Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-SHEEP IGG (GAMMA) AB HRP  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52200295
Affinity purified polyclonal secondary antibody against Sheep IgG, heavy chains (gamma) made in Rabbit and labeled with horseradish peroxidase. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-669-83