Learn More
anti-Sheep IgG, HRP, Isotype Control, Novus Biologicals™
Sheep Isotype Control
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP196876
Catalog No. NBP196876
Add to cart
Description
Description
IgG antibody specifically detects IgG in Sheep samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
Specifications
IgG
0.01 M Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2. Stabilizer: 10 mg/mL BSA - Immunoglobulin and Protease free. with No Preservative
immunoglobulin heavy constant gamma 1 (G1m marker)
Sheep
Ion exchange chromatography
RUO
Isotype Control
3500
Reconstitute with 1.0 ml deionized water (or equivalent) to desired concentration.
HRP
Lyophilized
IGHG1
IgG
1 mg
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Primary
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Peroxidase, anti- Sheep IgG and anti-Sheep Serum.
Sheep