Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sheep IgG Isotype Control, Rhodamine, Novus Biologicals™
Sheep Antibody
Supplier: Novus Biologicals NBP196911
Description
IgG specifically detects IgG in Sheep samples. It is validated for Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.Specifications
IgG | |
Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide | |
IGHG1 | |
Multi-step | |
RUO | |
3500 | |
Reconstitute with 1.0 ml deionized water (or equivalent). | |
Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Rhodamine | |
Immunoglobulin G, ImmunoglobulinG | |
Sheep | |
1 mg | |
Isotype Control (Primary) | |
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Sheep IgG and anti-Sheep Serum. | |
Sheep | |
Lyophilized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction