Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FBXW4P1 Monoclonal Antibody (2C5), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA521626
Description
Peptide Sequence: EVLLLHMCSY LDMRALGRLA QVYRWLWHFT NCDLLRRQIA WASLNSGFTR LGTNLMTSVP VKVSQNWIVG CCREGILLKW RCSQMPWMQL EDDALYISQA
FBXW4P1 Monoclonal antibody specifically detects FBXW4P1 in Human samples. It is validated for ELISA, Immunocytochemistry.Specifications
FBXW4P1 | |
Monoclonal | |
0.2-1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
0 | |
FBXW4P1 | |
FBXW4P1 (AAF04527, 31 a.a. ∽ 130 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Immunocytochemistry | |
2C5 | |
Unconjugated | |
FBXW4P1 | |
F-box and WD repeat domain containing 4 pseudogene 1; FBW3; FBXW3; FBXW4P1; SHFM3P1 | |
Mouse | |
Affinity chromatography | |
RUO | |
26226 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction