Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Shigella, Biotin, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA128922

Catalog No. PIPA128922

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA1-28922 detects Shigella from bacteria samples.PA1-28922 has been successfully used in immunofluorescence applications.The PA1-28922 immunogen is: Mixture of S. boydii, S. flexneri and S. dysenteriae.
Specifications

Specifications

Shigella
Polyclonal
Biotin
Purified
Mixture of S. boydii, S. flexneri and S. dysenteriae.
IgG fraction
RUO
Antibody
Polyclonal
Immunocytochemistry, Immunofluorescence
4mg/mL
PBS with 0.1% sodium azide; pH 7.2
Rabbit
IgG
100μL
Store at -20°C
Primary
Bacteria
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit