Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Shigella Rabbit anti-Bacteria, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA17245

Catalog No. PA17245


Add to cart

Description

Description

PA1-7245 detects Shigella sp. This antibody is polyvalent for Shigella antigens. The antiserum is unabsorbed and does cross-react with related Enterobacteriacea. PA1-7245 has been successfully used in ELISA and immunofluorescence procedures. The PA1-7245 immunogen is a mixture of Shigella boydii, Shigella flexneri and Shigella dysenteriae.

Specifications

Specifications

Shigella
Polyclonal
Unconjugated
Liquid
Mixture of Shigella boydii, Shigella flexneri and Shigella dysenteriae.
Purified
RUO
Antibody
Polyclonal
ELISA, Immunofluorescence
4-5 mg/mL
PBS with 0.1% sodium azide; pH 7.2
Rabbit
IgG
1 mL
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Primary
Bacteria
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit