Learn More
SHIP Mouse anti-Human, Mouse, Clone: P1C1, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 051911
Catalog No. 05-191-1MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This SHimmunoprecipitation antibody is validated for use in western blot, immunocytochemistry and immunoprecipitation for the detection of the SHimmunoprecipitation protein.
Specifications
Specifications
SHIP
P1C1
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Purified
Murine
IgG1, κ, κ
100μg
Primary
Human, Murine
Immunocytochemistry, Immunoprecipitation, Western Blot
Unconjugated
NP_001017915.1
INPP5D, hp51CN, p150Ship, SHIP, SIP-145, SHIP-1, SHIP1
KLH-conjugated linear peptide corresponding to human SHIP.
Protein G purfied
2-8°C for one year
Monoclonal