Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHISA2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525853

Catalog No. PIPA525853

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

SHISA2
Polyclonal
PBS with 0.09% sodium azide
Q6UWI4, Q8QZV2
C13orf13, PRO28631, TMEM46, WGAR9166, bA398O19.2, hShisa, protein shisa-2 homolog, transmembrane protein 46
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
219134, 387914
Immunohistochemistry, Western Blot
Unconjugated
SHISA2
Liquid
SHISA2
KLH conjugated synthetic peptide between 259-288 amino acids from the C-terminal region of human SHISA2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit