Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHISA9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA521058
Description
A suggested positive control is rat brain tissue lysate. PA5-21058 can be used with blocking peptide PEP-1172.
SHISA9 Polyclonal antibody specifically detects SHISA9 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
SHISA9 | |
Polyclonal | |
Unconjugated | |
SHISA9 | |
2700045P11Rik; Ckamp44; cystine-knot AMPAR modulating protein; cystine-knot AMPAR modulating protein of 44 kDa; Protein shisa-9; shisa family member 9; shisa homolog 9; shisa homolog 9-like; SHISA9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100361134, 72555, 729993 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
B4DS77, Q9CZN4 | |
SHISA9 | |
A 22 amino acid peptide near the center of human SHISA9 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction