Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

short stature homeobox 2, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human SHOX2 protein.

Manufacturer:  Abnova Corporation H00006474B01

Catalog No. 89-006-234


Add to cart

Description

Description

This gene is a member of the homeobox family of genes that encode proteins containing a 60-amino acid residue motif that represents a DNA binding domain. Homeobox genes have been characterized extensively as transcriptional regulators involved in pattern formation in both invertebrate and vertebrate species. Several human genetic disorders are caused by aberrations in human homeobox genes. This locus represents a pseudoautosomal homeobox gene that is thought to be responsible for idiopathic short stature, and it is implicated in the short stature phenotype of Turner syndrome patients. This gene is considered to be a candidate gene for Cornelia de Lange syndrome. Alternative splicing results in multiple transcript variants. [provided by RefSeq

Sequence: MEELTAFVSKSFDQKVKEKKEAITYREVLESGPLRGAKEPTGCTEAGRDDRSSPAVRAAGGGGGGGGGGGGGGGGGGVGGGGAGGGAGGGRSPVRELDMGAAERSREPGSPRLTEGRRKPTKAEVQATLLLPGEAFRFLVSPELKDRKEDAKGMEDEGQTKIKQRRSRTNFTLEQLNELERLFDETHYPDAFMREELSQRLGLSEARVQVWFQNRRAKCRKQENQLHKGVLIGAASQFEACRVAPYVNVGALRMPFQQDSHCNVTPLSFQVQAQLQLDSAVAHAHHHLHPHLAAHAPYMMFPAPPFGLPLATLAADSASAASVVAAAAAAKTTSKNSSIADLRLKAKKHAAALGL
Specifications

Specifications

short stature homeobox 2
Unconjugated
No additive
BC008829.2
OG12|OG12X|OGI2X|SHOT
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
6474
Western Blot
Mouse polyclonal antibody raised against a full-length human SHOX2 protein.
SHOX2
Serum
SHOX2
SHOX2 (AAH08829.1, 1 a.a. ~ 355 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit