Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHP-1/2 Rabbit anti-Human, Mouse, Clone: NL213, MilliporeSigma™
Rabbit Monoclonal Antibody
Supplier: MilliporeSigma 04742
Description
This Anti-SHP-1/2 Antibody, clone NL213 is validated for use in immunoprecipitation, western blotting for the detection of SHP-1/2.Specifications
SHP-1/2 | |
Monoclonal | |
Unconjugated | |
NM_002831, NM_002834 | |
Full-length glutathione transferase SHP-1 fusion protein, purified from bacteria | |
Primary | |
Human, Mouse | |
Supernatant |
Immunoprecipitation, Western Blot | |
NL213 | |
Cultured supernantant in 0.05% sodium azide | |
Rabbit | |
100 μL | |
PTPN6, HCP, SH-PTP1, PTP-1C, SHP1, SHP-1L, PTP1C, HCPH, SHP-1, HPTP1C | |
−20°C for 12 months. Aliquot to avoid freeze/thaw cycles. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only