Learn More
SHP-2/SHPTP-2 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 06118
Catalog No. 06-118-MI
Add to cart
Description
Description
Detect SHP-2/SHPTP-2 using this Anti-SHP-2/SHPTP-2 Antibody validated for use in IC and western blotting.
Specifications
Specifications
SHP-2/SHPTP-2
Unconjugated
Purified
Rabbit
IgG
400μg
Primary
NM_003179
Immunocytochemistry, Immunoprecipitation, Western Blot
Protein A Purified immunoglobulin in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105M NaCl, 0.035% sodium azide as a preservative.
PTPN11; PTP-2C; SH-PTP2; SHP-2; PTP2C; BPTP3; SHPTP2; SHP2; SH-PTP3; MGC14433; Shp2; PTP-1D; CFC; NS1
GST-fusion protein containing residues 2-216 of mouse SHP-2/SHPTP-2
Protein A purfied
−20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles.
Polyclonal
Human, Murine, Rat