Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CENPI Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA146129

Catalog No. PIPA146129


Add to cart

Description

Description

The immunogen shares 95% sequence identity to rat and 90% sequence identity to human.

A novel gene containing SNF2, PHD-finger, RING-finger, helicase, and linker histone domains termed SHPRH, is a possible candidate for a tumor suppressor gene. These domains are characteristic of several DNA repair proteins, transcription factors, and helicases. Mapping of this gene to 6q24, a region reported to contain a tumor suppressor locus suggests a role in tumor suppression.
Specifications

Specifications

CENPI
Polyclonal
Unconjugated
Cenpi
Liquid
CENPI
Residues 515-535 of the mouse SHPRH protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q8K1K4
2610103K11Rik, AA450458, AU024614, BC006883, D230017O13Rik, E130018M05
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
102920
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit