Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ST8SIA3 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA525508
Description
This antibody is predicted to react with mouse based on sequence homology.
ST8SIA3 belongs to a family of sialyltransferases that form sialyl-alpha-2,8-sialyl-R linkages at the nonreducing termini of glycoconjugates.Specifications
ST8SIA3 | |
Polyclonal | |
ST8SIA3 | |
SIAT8C, ST8SiaIII, SIAT8-C, alpha-2,8-sialyltransferase 8C, alpha-2,8-sialyltransferase III, sia-alpha-2,3-Gal-beta-1,4-GlcNAc-R:alpha 2,8-sialyltransferase, sialyltransferase 8C (alpha2,3Galbeta1,4GlcNAcalpha 2,8-sialyltransferase), sialytransferase St8Sia III | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51046 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
Unconjugated | |
O43173 | |
ST8SIA3 | |
KLH conjugated synthetic peptide between 312-340 amino acids from the C-terminal region of human SIA8C | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction