Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIGLEC12 Rat anti-Mouse, Clone: 750620, Invitrogen™

Rat Monoclonal Antibody

Manufacturer:  Invitrogen MA524359

Catalog No. PIMA524359


Add to cart

Description

Description

Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.

Sialic acid-binding immunoglobulin-like lectins (SIGLECs) are a family of cell surface proteins belonging to the immunoglobulin superfamily. They mediate protein-carbohydrate interactions by selectively binding to different sialic acid moieties present on glycolipids and glycoproteins. This gene encodes a member of the SIGLEC3-like subfamily of SIGLECs. Members of this subfamily are characterized by an extracellular V-set immunoglobulin-like domain followed by two C2-set immunoglobulin-like domains, and the cytoplasmic tyrosine-based motifs ITIM and SLAM-like. The encoded protein, upon tyrosine phosphorylation, has been shown to recruit the Src homology 2 domain-containing protein-tyrosine phosphatases SHP1 and SHP2. It has been suggested that the protein is involved in the negative regulation of macrophage signaling by functioning as an inhibitory receptor. This gene is located in a cluster with other SIGLEC3-like genes on 19q13.4. Alternatively spliced transcript variants encoding distinct isoforms have been described for this gene.
Specifications

Specifications

SIGLEC12
Monoclonal
0.2 mg/mL
PBS with 5% trehalose and No Preservative
Q91Y57
S2V, SIGLECL1, SLG, Siglec-XII, SIGLEC-like 1, sialic acid-binding Ig-like lectin 12, sialic acid-binding Ig-like lectin-like 1
Rat
IgG2B
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
83382
Flow Cytometry
750620
Unconjugated
Siglece
Lyophilized
Siglece
Mouse myeloma cell line NS0-derived recombit mouse Siglec-E Gln19-Phe354
Protein A/G
RUO
Antibody
Monoclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit