Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Siglec-3/CD33, Fluorescein, Clone: 6C5/2, R&D Systems™
Supplier: R&D Systems FAB1137F100
This item is not returnable.
View return policy
Description
Applications:
Flow Cytometry
Specifications
Siglec-3/CD33 | |
Monoclonal | |
Fluorescein | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD33 | |
Recombinant human Siglec-3/CD33, Extracellular domain | |
100 Tests | |
Primary | |
Detects human Siglec-3/CD33 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Siglec-2, -5, -7, or -9 is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG1 |
Flow Cytometry | |
6C5/2 | |
Flow Cytometry 10 uL/10^6 cells | |
CD33, CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, gp67, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, Siglec3, SIGLEC-3, SIGLEC3gp67 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
945 | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction