Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Siglec-3/CD33, PE, Clone: 6C5/2, R&D Systems™

Manufacturer:  R&D Systems FAB1137P100

 View more versions of this product

Catalog No. FAB1137P100


Add to cart

Description

Description

Applications:

Flow Cytometry

Specifications

Specifications

Siglec-3/CD33
6C5/2
Flow Cytometry 10 uL/10^6 cells
Purified
CD33
Recombinant human Siglec-3/CD33 Extracellular domain
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
945
Human
Flow Cytometry
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
CD33, CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, gp67, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, Siglec3, SIGLEC-3, SIGLEC3gp67
Mouse
IgG1
100 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human Siglec-3/CD33 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Siglec-2, -5, -7, or -9 is observed.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit