Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Siglec-3/CD33, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals 19170002

Catalog No. 19-170-002K


Add to cart

Description

Description

Siglec-3/CD33 Polyclonal antibody specifically detects Siglec-3/CD33 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Siglec-3/CD33
Unconjugated
20mM Potassium Phosphate (pH 7.0) and 0.15M NaCl with No Preservative
Affinity Purified
CD33
This antibody was made against a protein fragment from the N Terminus Region
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
This product is specific for Human CD33.
ELISA, Western Blot
Western Blot 1:5000-1:20000, ELISA 1:100-1:2000
NM_001772
CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, SIGLEC-3, SIGLEC3gp67
Rabbit
IgG
0.05 mg
Immunology
Primary
945
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only