Learn More
anti-Siglec-7/CD328, Alexa Fluor 488, Clone: 194211, R&D Systems™
Manufacturer:  R&D Systems FAB11381G100
 View more versions of this product
Catalog No. FAB11381G10
Add to cart
Description
Description
Applications:
Flow Cytometry
Specifications
Specifications
Siglec-7/CD328
194211
Flow Cytometry 5 uL/10^6 cells
Q9Y286
Adhesion inhibitory receptor molecule 1, adhesion inhibitory receptor molecule 1, siglec-7, AIRM1, AIRM-1, AIRM1QA79 membrane protein, CD328, CD328 antigen, CDw328, D-siglec, p75, p75/AIRM1, QA79, sialic acid binding Ig-like lectin 7, sialic acid binding immunoglobulin-like lectin 7, sialic acid-binding Ig-like lectin 7, Siglec7, Siglec-7
Mouse
IgG1
100 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects human Siglec-7 in ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Siglec-6, -8, -9 or recombinant Siglec-E is observed.
Flow Cytometry
Alexa Fluor 488
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
SIGLEC7
Mouse myeloma cell line NS0-derived recombinant human Siglec-7, Gln19-Gly357 (predicted), Accession # Q9Y286
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
27036
Human