Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-G Mouse anti-Mouse, PE-CF594, Clone: SH1, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 563337

Catalog No. BDB563337


Add to cart

Description

Description

The SH1 monoclonal antibody specifically binds to mouse Siglec-G (sialic acid binding immunoglobulin-like lectin G), also known as Siglec10. Siglec-G is a Type I transmembrane glycoprotein that belongs to the Immunoglobulin superfamily. Siglec-G functions as an adhesion molecule that mediates sialic-acid dependent binding to cells. Siglec-G is expressed on cells of the B cell lineage. Siglec-G is most highly expressed by pre-B cells and B1a cells within the B cell lineage and is not detectable on T cells. Siglec-G can inhibit B cell receptor-mediated calcium signaling when it is overexpressed. Mice that lack Siglec-G have significantly increased numbers of B1a cells that begins early in development and is B cell intrinsic. Siglec-G-deficient mice have higher titers of natural IgM antibodies than their normal counterparts. Mouse Siglec-G is the ortholog of human Siglec-10 (also known as, CD330).

This antibody is conjugated to BD Horizon™ PE-CF594, which has been developed exclusively by BD Biosciences as a better alternative to PE-Texas Red™. PE-CF594 excites and emits at similar wavelengths to PE-Texas Red™ yet exhibits improved brightness and spectral characteristics. Due to PE having maximal absorption peaks at 496nm and 564nm, PE-CF594 can be excited by the blue (488-nm), green (532-nm) and yellow-green (561-nm) lasers and can be detected with the same filter set as PE-Texas Red™ (eg 610/20-nm filter).

Specifications

Specifications

Siglec-G
SH1
PE-CF594
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
IgG1, κ
RUO
Primary
Murine
Flow Cytometry
0.2mg/mL
Siglecg; mSiglec-G; Siglec10; Siglec 10; CD330
Affinity Purified
Mouse Extracellular Siglec-G Recombinant Protein
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit