Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIII p15 Mouse, Unlabeled, Clone: 56, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610760

 View more versions of this product

Catalog No. BDB610760


Add to cart

Description

Description

SIII p15 is one of three polypeptides that comprise the approximate 140kDa RNA polymerase II transcription factor SIII. The other two polypeptides have the molecular weights of 110kDa and 18kDa. All three polypeptides are required for transcription factor SIII function. Although the polypeptides were identified and purified from rat liver nuclear extracts, tissue expression of this transcription factor appears to be widespread. RNA polymerase II recognizes the SIII transcription factor and in turn initiates transcription from these correctly initiated transcripts. A yeast transcription factor designated YES may be the yeast homolog to SIII.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

SIII p15
56
Unconjugated
Affinity Purified
Human SIII p15 aa. 1-112
50μg
Cell Biology
Primary
Canine, Frog, Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.