Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIKE Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525292

Catalog No. PIPA525292


Add to cart

Description

Description

This antibody is predicted to react with bovine and rat based on sequence homology.

SIKE interacts with IKK-epsilon (IKBKE; MIM 605048) and TBK1 (MIM 604834) and acts as a suppressor of TLR3 (MIM 603029) and virus-triggered interferon activation pathways.
Specifications

Specifications

SIKE
Polyclonal
PBS with 0.09% sodium azide
Q9BRV8
SIKE, suppressor of IKK epsilon, suppressor of IKK-epsilon
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
80143
Immunohistochemistry, Western Blot
Unconjugated
SIKE1
Liquid
SIKE1
KLH conjugated synthetic peptide between 154-182 amino acids from the C-terminal region of human SIKE
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit