Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sin1/MAPKAP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$204.00 - $409.50
Specifications
Antigen | Sin1/MAPKAP1 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224523SS
|
Novus Biologicals
NBP224523SS |
0.025 mg |
Each for $204.00
|
|
|||||
NBP224523
|
Novus Biologicals
NBP224523 |
0.1 mg |
Each for $409.50
|
|
|||||
Description
Sin1/MAPKAP1 Polyclonal specifically detects Sin1/MAPKAP1 in Human, Mouse, Rat, Bovine, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Sin1/MAPKAP1 | |
Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml | |
Polyclonal | |
Rabbit | |
Autophagy, Cancer, mTOR Pathway, Protein Kinase | |
0.2 ml PBS and 0.2% Gelatin with 0.05% Sodium Azide | |
JC310, MEKK2-interacting protein 1, MGC2745, MIP1SIN1SIN1b, Mitogen-activated protein kinase 2-associated protein 1, mitogen-activated protein kinase associated protein 1, mSIN1, ras inhibitor MGC2745, SAPK-interacting protein 1, SIN1g, stress-activated map kinase interacting protein 1, Stress-activated map kinase-interacting protein 1, stress-activated protein kinase-interacting 1, target of rapamycin complex 2 subunit MAPKAP1, TORC2 subunit MAPKAP1 | |
MAPKAP1 | |
IgG | |
Affinity Purified | |
The amino acid sequence used as immunogen for the antibody is 100% homologous in human and cow, 94% homologous in mouse, rat and dog, 86% homologous in xenopus and chicken. |
0.5 mg/mL | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse, Rat, Bovine, Canine | |
Q92973 | |
79109 | |
A portion of amino acids 100-150 of human SIN1 was used as the immunogen for this antibody. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title