Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIRPB1 Monoclonal Antibody (308906), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA524026
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
SIRPB1 Monoclonal antibody specifically detects SIRPB1 in Human samples. It is validated for Flow Cytometry, Western Blot.Specifications
SIRPB1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
O00241 | |
SIRPB1 | |
Mouse myeloma cell line NS0-derived recombinant human SIRP beta 1/CD172b Gly26-Ala369 (Arg53His and Ala363Pro) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry, Western Blot | |
308906 | |
Unconjugated | |
SIRPB1 | |
9930027N05Rik; CD172 antigen-like family member B; CD172b; CD172b antigen; DKFZp686A05192; signal regulatory protein beta 1; signal-regulatory protein beta 1; signal-regulatory protein beta 1A; signal-regulatory protein beta-1; Signal-regulatory protein beta-1 isoform 3; SIRP BETA 1; SIRP beta 1 cell surface protein; Sirpb; SIRPB1; Sirpb1a; SIRP-beta; SIRP-beta B; SIRP-BETA-1; SIRP-beta-1 isoform 3 | |
Mouse | |
Protein A/G | |
RUO | |
10326 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction