Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIRT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA3200

Catalog No. PA3-200


Add to cart

Description

Description

PA3-200 detects human and rat SIRT2 and shows no cross-reactivity to other SIRT isoforms. PA3-200 has been successfully used in Western blot and immunocytochemistry procedures. By Western blot, this antibody detects an ~48 kDa protein representing SIRT2 from SAOS2 cells. PA3-200 can be used for immunocytochemistry procedures. PA3-200 immunizing peptide corresponds to amino acid residues 341-352 from the human SIRT2 protein. This sequence is completely conserved between human and rat.

The SIRT2 protein (also known as Sirtuin for Silent Mating Type Information 2-Homolog) is a NAD-dependent deacytylase (NDAC) that has been shown to control gene silencing, cell cycle, and DNA damage repair. It is believed that SIRT2 may act as a tumor suppressor in human gliomas and may also serve as a novel molecular marker for these cells. SIRT2 has also been shown to act as a redox sensor to help regulate muscle gene expression in response to food intake and exercise. SIRT2 acts in the phosphorylation cascade involving mitosis where SIRT2 is phosphorylated in late G2 phase, during M phase, and into cytokinesis.
Specifications

Specifications

SIRT2
Polyclonal
whole serum with 0.05% sodium azide
Q8IXJ6, Q8VDQ8, Q5RJQ4
Sirtuin for Silent Mating Type Information 2-Homolog
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
22933, 361532, 64383
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SIRT2
Liquid
SIRT2
Synthetic Peptide: D(341) E A R T T E R E K P Q(352)
NA
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit