Learn More
sirtuin (silent mating type information regulation 2 homolog) 3 (S. cerevisiae) (B02), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human SIRT3 protein.
Supplier: Abnova Corporation H00023410B02
Description
This gene encodes a member of the sirtuin family of proteins, homologs to the yeast Sir2 protein. Members of the sirtuin family are characterized by a sirtuin core domain and grouped into four classes. The functions of human sirtuins have not yet been determined; however, yeast sirtuin proteins are known to regulate epigenetic gene silencing and suppress recombination of rDNA. Studies suggest that the human sirtuins may function as intracellular regulatory proteins with mono-ADP-ribosyltransferase activity. The protein encoded by this gene is included in class I of the sirtuin family. Two alternatively spliced transcript variants that encode different proteins have been described for this gene. [provided by RefSeq
Sequence: MVGAGISTPSGIPDFRSPGSGLYSNLQQYDLPYPEAIFELPFFFHNPKPFFTLAKELYPGNYKPNVTHYFLRLLHDKGLLLRLYTQNIDGLERVSGIPASKLVEAHGTFASATCTVCQRPFPGEDIRADVMADRVPRCPVCTGVVKPDIVFFGEPLPQRFLLHVVDFPMADLLLILGTSLEVEPFASLTEAVRSSVPRLLINRDLVGPLAWHPRSRDVAQLGDVVHGVESLVELLGWTEEMRDLVQRETGKLDGPDKSpecifications
sirtuin (silent mating type information regulation 2 homolog) 3 (S. cerevisiae) | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human SIRT3 protein. | |
SIRT3 | |
SIR2L3 | |
Mouse | |
50 μL | |
Yes | |
23410 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
No additive | |
NM_001017524.1 | |
SIRT3 | |
SIRT3 (NP_001017524.1, 1 a.a. ∼ 257 a.a) full-length human protein. | |
RUO | |
Primary | |
Human | |
Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.