Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

anti-SIRT6, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP130101

Catalog No. NBP130101

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifications

Specifications

SIRT6
1.0 mg/mL
Western Blot 1:1000-1:5000, Immunoprecipitation 2-5 ug/mg lysate
Q8N6T7
EC 3.5.1.-, NAD-dependent deacetylase sirtuin-6, SIR2L6sir2-related protein type 6, SIR2-like protein 6, sirtuin (silent mating type information regulation 2 homolog) 6 (S. cerevisiae), sirtuin (silent mating type information regulation 2, S. cerevisiae, homolog) 6, sirtuin 6, sirtuin type 6
Rabbit
IgG
100 ul
Cancer, Chromatin Research, DNA Repair, Epigenetics, Histone Deacetylases
Primary
51548
Immunoprecipitation, Western Blot
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
SIRT6
Synthetic peptide mapping to a region between residue 305 and 355 of human sirtuin (silent mating type information regulation 2 homolog) 6 using the numbering given in entry NP_057623.1 (GeneID 51548).
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only