Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIRT6 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA513225

Catalog No. PIPA513225


Add to cart

Description

Description

SIRT6 is a member of the sirtuin family of proteins, homologs to the yeast Sir2 protein. Members of the sirtuin family are characterized by a sirtuin core domain and grouped into four classes. The functions of human sirtuins have not yet been determined; however, yeast sirtuin proteins are known to regulate epigenetic gene silencing and suppress recombination of rDNA. Studies suggest that the human sirtuins may function as intracellular regulatory proteins with mono-ADP-ribosyltransferase activity.
Specifications

Specifications

SIRT6
Polyclonal
PBS with 0.09% sodium azide
Q8N6T7, P59941
Mono-ADP-ribosyltransferase sirtuin-6; SIR2-like protein 6; SIR2L6
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
50721, 51548
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SIRT6
Liquid
SIRT6
KLH conjugated synthetic peptide between 290-319 amino acids from the C-terminal region of human SIRT6
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit