Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIRT7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA141029
Description
Suggested positive control: human liver or PBMC.
SIRT7 Polyclonal antibody specifically detects SIRT7 in Human samples. It is validated for Western Blot.Specifications
SIRT7 | |
Polyclonal | |
Unconjugated | |
SIRT7 | |
MGC126840; MGC126842; NAD-dependent deacetylase sirtuin-7; NAD-dependent protein deacetylase sirtuin-7; NAD-dependent protein deacylase sirtuin-7; regulatory protein SIR2 homolog 7; silent mating type information regulation 2, S.cerevisiae, homolog 7; Sir2l7; SIR2-like protein 7; sir2-related protein type 7; Sirt7; sirtuin 7; sirtuin type 7; Unknown (protein for MGC:142712) | |
Rabbit | |
Protein G | |
RUO | |
51547 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1.0 mg/mL | |
PBS with 0.05% sodium azide | |
Q9NRC8 | |
SIRT7 | |
Synthetic peptides corresponding to residues 35-51 and 361-377 of human SIRT7. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction