Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Sirtuin 1/SIRT1, Alexa Fluor 488, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP149540AF488

Catalog No. NBP149540X

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Sirtuin 1/SIRT1
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
Affinity Purified
SIRT1
A genomic peptide made to an internal region of the human SIRT1 protein (within residues 400-550). [Swiss-Prot Q96EB6]
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Q96EB6
EC 3.5.1, S. cerevisiae, homolog) 1, sirtuin 1
Rabbit
IgG
0.1 ml
Apoptosis, Chromatin Research, DNA Repair, Epigenetics, Histone Deacetylases, Phospho Specific
Primary
23411
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only