Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SKAP55 Mouse, Unlabeled, Clone: 35, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611236

Catalog No. BDB611236


Add to cart

Description

Description

Immediately following T cell receptor (TcR) ligation, cytoplasmic protein-tyrosine kinases (PTKs) are activated, resulting in the phosphorylation of intracellular and transmembrane proteins. p56[lck] and p59[fyn] are two Src family PTKs activated by TcR ligation. Following activation, Fyn interacts with several tyrosine-phosphorylated proteins, including SKAP55 (Src kinase-associated phosphoprotein of 55kDa). SKAP55, which selectively binds Src kinase SH2 domains, contains a pleckstrin homology (PH) domain, a C-terminal Src homology 3 (SH3) domain, and multiple tyrosine phosphorylation sites. In addition, SKAP55 directly binds to FYB/SLAP-130, a novel substrate of TcR-stimulated protein tyrosine kinases. Like SLAP-130, SKAP55 is expressed exclusively in mononuclear cells, with preferential expression in T lymphocytes where it is constitutively phosphorylated in resting cells. A SKAP55 homolog, SKAP-HOM, has been identified which also interacts with SLAP-130, exhibits ubiquitous expression, and is phosphorylated only following T cell activation. Therefore, the Fyn-associated protein, SKAP55, along with SLAP-130, are components of a Fyn-mediated signaling cascade in T cells.

Host Species: Mouse
Clone: 35
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Human SKAP55 aa. 154-353
Formula Weight [Chemical]: 55kDa

Immunofluorescence, Western Blotting

Specifications

Specifications

SKAP55
35
Unconjugated
Affinity Purified
Human SKAP55 aa. 154-353
50μg
Cell Biology
Primary
Human, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit