Learn More
SKAP55 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA1525
Catalog No. PA1-525
Add to cart
Description
Description
PA1-525 detects human SKAP55. PA1-525 has been successfully used in Western blot, immunocytochemistry and immunoprecipitation procedures. By Western blot, this antibody detects an ~ 60 kDa protein from human Jurkat cells.This antibody has also been used in immunoprecipitation and immunocytochemical procedures. PA1-525 antigen corresponds to amino acids 2-105 of human SKAP-55.
Src kinase-associated phosphoprotein of 55 kDa (SKAP55) is expressed as a constitutively tyrosine phosphorylated 55-kD adapter protein that positively regulates TCR signaling. SKAP55 has also been shown to enhance adhesion to fibronectin and ICAM1, colocalize with F-actin at the T cell-antigen-presenting cell synapse and promotes the clustering of LFA1 and TCR ligation. TCR engagement induces the translocation of SKAP55 to lipid rafts, where the interactions with Fyn take place. CD45 also plays a very important role in SKAP55/TCR-mediated signaling. SKAP55 involvement in the TCR signaling pathway is the subject of current research in murine as well as human models.
Specifications
Specifications
SKAP55
Polyclonal
Unconjugated
SKAP1
Liquid
SKAP1
Immunogen corresponds to amino acids 2-105 from human SKAP55.
Protein G
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunoprecipitation, Western Blot
1 mg/mL
PBS with 1mg/mL BSA and 0.02% sodium azide
Q86WV1
SCAP1, SKAP55 adaptor protein, SRC family associated phosphoprotein 1, SRC kinase associated phosphoprotein 55 kD
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
8631