Learn More
Brady™ Anti-Skid Tape Roll - Grit-Coated Polyester, Solid Color, Red
Grit-Coated Polyester Tape (B-916) from Brady™ is specifically formulated to provide traction on steps, slick floors and egress pathways.
$31.50 - $120.00
Specifications
Product Type Anti-Skid Tape
View More Specs
Products 3
Catalog Number Mfr. No. Width (English) Price Quantity & Availability  
Catalog Number Mfr. No. Width (English) Price Quantity & Availability  
19-103-793 View Documents Brady™
78197
1 in. Each for $31.50
Add to cart
19-103-794 View Documents Brady™
78198
2 in. Each for $60.50
Add to cart
19-103-795 View Documents Brady™
78199
4 in. Each for $120.00
Add to cart
Description
Description
  • This indoor/outdoor anti-skid tape has a permanent pressure sensitive acrylic adhesive
  • Excellent chemical, oil and water resistance
  • Anti-skid tape helps prevent slips, trips and falls in wet or high traffic areas
  • Performs in temperatures ranging from -40°F to 158°F (-40°C to 70°C)
  • Apply at 50°F (10°C) or higher
Specifications
Specifications
Anti-Skid Tape