Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SKI-like oncogene, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human SKIL protein.

Manufacturer:  Abnova Corporation H00006498B01

Catalog No. 89-006-239


Add to cart

Description

Description

Sequence: MENLQTNFSLVQGSTKKLNGMGDDGSPPAKKMITDIHVNGKTINKVPTVKKEHLDDYGEAPVETDGEHVKRTCTSVPETLHLNPSLKHTLAQFHLSSQSSLGGPAAFSARHSQESMSPTVFLPLPSPQVLPGPLLIPSDSSTELTQTVLEGESISCFQVGGEKRLCLPQVLNSVLREFTLQQINTVCDELYIYCSRCTSDQLHILKVLGILPFNAPSCGLITLTDAQRLCNALLRPRTFPQNGSVLPAKSSLAQLKETGSAFEVEHECLGKCQGLFAPQFYVQPDAPCIQCLECCGMFAPQTFVMHSHRSPDKRTCHWGFESAKWHCYLHVNQKYLGTPEEKKLKIILEEMKEKFSMRSGKRNQSKTDAPSGMELQSWYPVIKQEGDHVSQTHSFLHPSYYLYMCDKVVAPNVSLTSAVSQSKELTKTEASKSISRQSEKAHSSGKLQKTVSYPDVSLEEQEKMDLKTSRELCSRLDASISNNSTSKRKSESATCNLVRDINKVGIGLVAAASSPLLVKDVICEDDKGKIMEEVMRTYLKQQEKLNLILQKKQQLQMEVKMLSSSKSMKELTEEQQNLQKELESLQNEHAQRMEEFYVEQKDLEKKLEQIMKQKCTCDSNLEKDKEAEYAGQLAELRQRLDHAEADRQELQDELRQEREARQKLEMMIKELKLQILKSSKTAKE
Specifications

Specifications

SKI-like oncogene
Unconjugated
No additive
NM_005414.2
SNO|SnoA|SnoN
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
6498
Western Blot
Mouse polyclonal antibody raised against a full-length human SKIL protein.
SKIL
Serum
SKIL
SKIL (NP_005405.1, 1 a.a. ~ 684 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit