Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Slap, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP224696

 View more versions of this product

Catalog No. NBP224696


Add to cart

Description

Description

Slap Polyclonal antibody specifically detects Slap in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications

Specifications

Slap
0.5 mg/mL
Immunohistochemistry-Paraffin 5-15 ug/ml
Purified
SLA
A portion of amino acids 175-225 of human SLAP was used as the immunogen for the antibody.
Protein A purified
RUO
Primary
6503
Human
Immunohistochemistry (Paraffin)
Unconjugated
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
hSLAP, SLA1, SLAP, SLAP1, SLAP-1, src-like-adapter, Src-like-adapter protein 1, Src-like-adaptor
Rabbit
IgG
0.1 mg
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
The UNC5CL amino acid sequence used as immunogen for the antibody is 100% homologous in human and chimpanzee and 88% homologous in rhesus monkey.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only