Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC10A4 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523755

Catalog No. PIPA523755


Add to cart

Specifications

Specifications

SLC10A4
Polyclonal
PBS with 0.09% sodium azide
Q96EP9, Q3UEZ8
P4, Na(+)/bile acid cotransporter 4, bile acid transporter SLC10A4, sodium/bile acid cotransporter 4, solute carrier family 10 member 4
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
201780, 231290
Western Blot
Unconjugated
SLC10A4
Liquid
SLC10A4
KLH conjugated synthetic peptide between 370-398 amino acids from the C-terminal region of human SLC10A4
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit