Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC16A9 Rabbit anti-Hamster, Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA525827
Description
Specifications
SLC16A9 | |
Polyclonal | |
Unconjugated | |
Q7RTY1 | |
SLC16A9 | |
KLH conjugated synthetic peptide between 194-223 amino acids from the central region of human SLC16A9 | |
400 μL | |
Primary | |
Hamster, Human | |
Antibody | |
IgG |
Western Blot | |
0.5 mg/mL | |
SLC16A9 | |
C10orf36, MCT9, MCT 9, monocarboxylate transporter 9, solute carrier family 16 (monocarboxylic acid transporters), member 9, solute carrier family 16 member 9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100760512, 220963 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction