Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASCT2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526445

Catalog No. PIPA526445


Add to cart

Description

Description

SLC1A5 (Solute carrier family 1 (neutral amino acid transporter), member 5) is a member of the Na(+)-dependent amino acid transporter superfamily. It has a broad substrate specificity, a preference for zwitterionic amino acids, and a sodium-dependence. It accepts as substrates all neutral amino acids, including glutamine, asparagine, and branched-chain and aromatic amino acids, and excludes methylated amino acids, anionic amino acids, and cationic amino acids. It acts as a cell surface receptor for feline endogenous virus RD114, baboon M7 endogenous virus and type D simian retroviruses.
Specifications

Specifications

ASCT2
Polyclonal
PBS with 0.09% sodium azide
Q15758
AAAT, ASCT2, ATBO, M7V1, M7VS1, R16, RDRC, ATB(0), RD114 virus receptor, RD114/simian type D retrovirus receptor, baboon M7 virus receptor, neutral amino acid transporter B, neutral amino acid transporter B(0), sodium-dependent neutral amino acid transporter type 2, solute carrier family 1 member 5
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6510
Western Blot
Unconjugated
SLC1A5
Liquid
SLC1A5
KLH conjugated synthetic peptide between 186-214 amino acids from the central region of human SLC1A5
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit