Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC25A12 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523778

Catalog No. PIPA523778

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine, mouse, rat and Xenopus based on sequence homology.

This gene encodes a calcium-binding mitochondrial carrier protein. The encoded protein localizes to the mitochondria and is involved in the exchange of aspartate for glutamate across the inner mitochondrial membrane. Polymorphisms in the gene encoding this protein may be associated with risk of autism.
Specifications

Specifications

SLC25A12
Polyclonal
PBS with 0.09% sodium azide
O75746, Q8BH59
AGC1, ARALAR, araceli hiperlarga, calcium binding mitochondrial carrier superfamily member Aralar1, calcium-binding mitochondrial carrier protein Aralar1, mitochondrial aspartate glutamate carrier 1, solute carrier family 25 member 12
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
78830, 8604
Immunohistochemistry, Western Blot
Unconjugated
SLC25A12
Liquid
SLC25A12
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human SLC25A12
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit