Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC25A19 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525193

Catalog No. PIPA525193


Add to cart

Description

Description

This antibody is predicted to react with non-human primate based on sequence homology.

SLC25A19 encodes a mitochondrial protein that is a member of the solute carrier family. Although this protein was initially thought to be the mitochondrial deoxynucleotide carrier involved in the uptake of deoxynucleotides into the matrix of the mitochondria, further studies have demonstrated that this protein instead functions as the mitochondrial thiamine pyrophosphate carrier, which transports thiamine pyrophosphates into mitochondria.
Specifications

Specifications

SLC25A19
Polyclonal
PBS with 0.09% sodium azide
Q9HC21, Q9DAM5
DNC, MCPHA, MUP1, TPC, mitochondrial thiamine pyrophosphate carrier, mitochondrial uncoupling protein 1, solute carrier family 25 (mitochondrial deoxynucleotide carrier), member 19
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
60386, 67283
Western Blot
Unconjugated
SLC25A19
Liquid
SLC25A19
KLH conjugated synthetic peptide between 236-263 amino acids from the C-terminal region of human SLC25A19
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit